Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080024.2 | NP_001073493.2 | 5 - 6 | 1496 | 69 | |
GenBank mRNA | BC124720.1 | - | 5 - 6 | 1496 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |