Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b4galnt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080570.1 | NP_001074039.1 | 4 - 5 | 550 | 121 | |
XM_005162256.4 | XP_005162313.1 | 4 - 5 | 651 | 121 | ||
XM_021469816.1 | XP_021325491.1 | 4 - 5 | 644 | 121 | ||
GenBank mRNA | BC129323.1 | - | 4 - 5 | 550 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |