Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gli2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130967.1 | NP_571042.1 | 13 - 14 | 2565 | 81 | |
GenBank mRNA | AF085746.2 | - | 13 - 14 | 2565 | 81 | |
AF348156.1 | - | 10 - 11 | 1956 | 81 | ||
BC163542.1 | - | 13 - 14 | 2556 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |