Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131357.1 | NP_571432.1 | 5 - 6 | 661 | 68 | |
GenBank mRNA | AF132084.1 | - | 5 - 6 | 661 | 68 | |
BC162211.1 | - | 5 - 6 | 603 | 68 | ||
BC162544.1 | - | 5 - 6 | 603 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |