Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131407.1 | NP_571482.1 | 6 - 7 | 613 | 80 | |
GenBank mRNA | BC085415.1 | - | 6 - 7 | 613 | 80 | |
BC165821.1 | - | 6 - 7 | 531 | 80 | ||
DQ360071.1 | - | 6 - 7 | 497 | 80 | ||
Y15075.1 | - | 581 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |