Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mhc1uba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131471.1 | NP_571546.1 | 7 - 8 | 1126 | 72 | |
GenBank mRNA | BC074095.1 | - | 7 - 8 | 1100 | 72 | |
Z46777.1 | - | 1126 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |