Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfrsfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131840.2 | NP_571915.2 | 4 - 5 | 667 | 60 | |
GenBank mRNA | AF250042.2 | - | 4 - 5 | 597 | 60 | |
BC162652.1 | - | 4 - 5 | 567 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |