Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnf1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131880.3 | NP_571955.3 | 2 - 3 | 700 | 59 | |
XM_021481477.1 | XP_021337152.1 | 2 - 3 | 705 | 59 | ||
XM_021481478.1 | XP_021337153.1 | 2 - 3 | 409 | 59 | ||
GenBank mRNA | AF244140.2 | - | 2 - 3 | 643 | 59 | |
AF430840.1 | - | 2 - 3 | 707 | 59 | ||
AY336602.1 | - | 2 - 3 | 614 | 59 | ||
AY569309.1 | - | 2 - 3 | 666 | 59 | ||
BC129147.1 | - | 2 - 3 | 675 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |