Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnbpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244736.1 | NP_001231665.1 | 5 | 670 | 150 | |
NM_199749.2 | NP_956043.2 | 5 | 763 | 150 | ||
XM_005170569.3 | XP_005170626.1 | 5 | 716 | 150 | ||
GenBank mRNA | BC056793.1 | - | 5 | 763 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |