Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312897.1 | NP_001299826.1 | 12 - 13 | 2143 | 140 | |
NR_132439.1 | 14 - 15 | 2223 | 140 | |||
GenBank mRNA | BC047798.1 | - | 14 - 15 | 2217 | 140 | |
BC057484.1 | - | 11 - 12 | 1481 | 140 | ||
EE314390.1 | - | 5 - 6 | 601 | 140 | ||
EG573628.1 | - | 7 - 8 | 709 | 140 |
Gene Symbol: LOC110437844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021471075.1 | XP_021326750.1 | 5 - 6 | 580 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |