Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312897.1 | NP_001299826.1 | 7 - 8 | 1587 | 133 | |
NR_132439.1 | 8 - 9 | 1457 | 133 | |||
GenBank mRNA | BC047798.1 | - | 8 - 9 | 1451 | 133 | |
EE718067.1 | - | 1 - 2 | 145 | 133 | ||
HM151010.1 | - | 7 - 8 | 1390 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |