Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199836.2 | NP_956130.1 | 6 - 7 | 724 | 83 | |
GenBank mRNA | BC044149.1 | - | 6 - 7 | 725 | 83 | |
BC163944.1 | - | 6 - 7 | 568 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |