Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200179.1 | NP_956473.1 | 14 - 15 | 1506 | 67 | |
XM_005166673.4 | XP_005166730.1 | 14 - 15 | 1594 | 67 | ||
GenBank mRNA | BC045476.1 | - | 14 - 15 | 1506 | 67 | |
BC165560.1 | - | 12 - 13 | 1209 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |