Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: maco1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200463.2 | NP_956757.2 | 9 - 10 | 1845 | 108 | |
XM_005157048.4 | XP_005157105.1 | 9 - 10 | 1821 | 108 | ||
XM_021480543.1 | XP_021336218.1 | 9 - 10 | 1926 | 108 | ||
GenBank mRNA | AY845030.1 | - | 9 - 10 | 1845 | 108 | |
BC171392.1 | - | 9 - 10 | 1815 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |