Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agfg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200588.2 | NP_956882.2 | 10 - 11 | 1324 | 100 | |
XM_005163316.4 | XP_005163373.1 | 9 - 10 | 1302 | 100 | ||
GenBank mRNA | BC056733.1 | - | 10 - 11 | 1324 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |