Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trappc6bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200661.1 | NP_956955.1 | 3 - 4 | 193 | 141 | |
GenBank mRNA | BC057509.1 | - | 3 - 4 | 193 | 141 | |
BC068383.1 | - | 3 - 4 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |