Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trappc6bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200661.1 | NP_956955.1 | 5 - 6 | 394 | 119 | |
GenBank mRNA | BC057509.1 | - | 5 - 6 | 394 | 119 | |
BC068383.1 | - | 5 - 6 | 436 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |