Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201071.1 | NP_957365.1 | 3 - 4 | 408 | 128 | |
GenBank mRNA | BC046019.1 | - | 3 - 4 | 408 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |