Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhot1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212704.1 | NP_997869.1 | 13 - 14 | 1406 | 69 | |
XM_009299425.3 | XP_009297700.1 | 13 - 14 | 1435 | 69 | ||
XM_009299426.3 | XP_009297701.1 | 13 - 14 | 1434 | 69 | ||
XM_009299427.3 | XP_009297702.1 | 13 - 14 | 1435 | 69 | ||
GenBank mRNA | BC044431.1 | - | 13 - 14 | 1401 | 69 | |
BC068190.1 | - | 13 - 14 | 1406 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |