Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agxtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213162.1 | NP_998327.1 | 2 - 3 | 458 | 62 | |
GenBank mRNA | BC056520.1 | - | 2 - 3 | 458 | 62 | |
BC067638.1 | - | 2 - 3 | 444 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |