Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002593.2 | NP_001002593.1 | 4 - 5 | 501 | 142 | |
GenBank mRNA | BC076005.1 | - | 4 - 5 | 452 | 142 | |
EH448113.1 | - | 4 - 5 | 494 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |