Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007415.1 | NP_001007416.1 | 17 - 18 | 1858 | 84 | |
XM_005171163.4 | XP_005171220.1 | 17 - 18 | 1719 | 84 | ||
XM_021480037.1 | XP_021335712.1 | 17 - 18 | 1837 | 84 | ||
GenBank mRNA | BC085370.1 | - | 17 - 18 | 1858 | 84 | |
BC165850.1 | - | 16 - 17 | 1637 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |