Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014289.2 | NP_001014311.1 | 10 - 11 | 1213 | 144 | |
GenBank mRNA | AB096100.1 | - | 10 - 11 | 1279 | 144 | |
BC059195.1 | - | 10 - 11 | 1212 | 144 | ||
BC165852.1 | - | 3 - 4 | 408 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |