Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014289.2 | NP_001014311.1 | 18 - 19 | 2510 | 120 | |
GenBank mRNA | AB096100.1 | - | 18 - 19 | 2576 | 120 | |
BC059195.1 | - | 18 - 19 | 119 | |||
BC165852.1 | - | 11 - 12 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |