Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ets1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017558.1 | NP_001017558.1 | 3 - 4 | 558 | 110 | |
GenBank mRNA | BC092935.1 | - | 3 - 4 | 558 | 110 | |
BC165216.1 | - | 417 | 110 | |||
KF774190.1 | - | 3 - 4 | 511 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |