Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039679.1 | NP_001034768.1 | 4 - 5 | 534 | 103 | |
GenBank mRNA | AY309263.1 | - | 4 - 5 | 534 | 103 | |
BC162727.1 | - | 4 - 5 | 565 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |