Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040687.1 | NP_001035777.1 | 25 - 26 | 3284 | 131 | |
XM_017351014.2 | XP_017206503.1 | 27 - 28 | 3876 | 131 | ||
XM_021481465.1 | XP_021337140.1 | 26 - 27 | 3760 | 131 | ||
GenBank mRNA | DQ466904.1 | - | 25 - 26 | 3284 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |