Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcnh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042722.2 | NP_001036187.2 | 8 - 9 | 2610 | 82 | |
XM_021480879.1 | XP_021336554.1 | 8 - 9 | 2585 | 82 | ||
XM_021480881.1 | XP_021336556.1 | 4 - 5 | 2014 | 82 | ||
GenBank mRNA | DQ022613.1 | - | 8 - 9 | 2150 | 82 | |
HM209079.1 | - | 8 - 9 | 2150 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |