Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328211.1 | NP_001315140.1 | 10 - 11 | 1820 | 66 | |
XM_005169952.4 | XP_005170009.1 | 10 - 11 | 2017 | 66 | ||
XM_009293507.3 | XP_009291782.1 | 10 - 11 | 2014 | 66 | ||
GenBank mRNA | BC124825.1 | - | 4 - 5 | 426 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |