Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328211.1 | NP_001315140.1 | 12 - 13 | 2107 | 64 | |
XM_005169952.4 | XP_005170009.1 | 12 - 13 | 2304 | 64 | ||
XM_009293507.3 | XP_009291782.1 | 12 - 13 | 2301 | 64 | ||
GenBank mRNA | BC124825.1 | - | 6 - 7 | 713 | 64 | |
BC134844.1 | - | 7 - 8 | 854 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |