Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shdb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045077.2 | NP_001038542.1 | 2 - 3 | 488 | 152 | |
XM_005170476.4 | XP_005170533.1 | 3 - 4 | 935 | 152 | ||
XM_009296270.3 | XP_009294545.1 | 3 - 4 | 936 | 152 | ||
XM_021469342.1 | XP_021325017.1 | 3 - 4 | 993 | 152 | ||
GenBank mRNA | BC097159.1 | - | 3 - 4 | 835 | 152 | |
BC122203.1 | - | 3 - 4 | 915 | 152 | ||
BC124504.1 | - | 3 - 4 | 909 | 152 | ||
BC129459.1 | - | 3 - 4 | 927 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |