Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crtac1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080178.1 | NP_001073647.1 | 12 - 13 | 1653 | 63 | |
XM_021475068.1 | XP_021330743.1 | 12 - 13 | 1734 | 63 | ||
GenBank mRNA | BC146702.1 | - | 12 - 13 | 63 | ||
DQ351708.1 | - | 12 - 13 | 1653 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |