Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zcchc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098388.2 | NP_001091858.1 | 3 - 4 | 840 | 77 | |
GenBank mRNA | BC141714.1 | - | 3 - 4 | 769 | 77 | |
CT621901.2 | - | 3 - 4 | 390 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |