Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sema4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131049.1 | NP_571124.1 | 5 - 6 | 866 | 100 | |
GenBank mRNA | AF073289.1 | - | 5 - 6 | 866 | 100 | |
BC078280.1 | - | 4 - 5 | 471 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |