Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131191.1 | NP_571266.1 | 7 - 8 | 859 | 63 | |
GenBank mRNA | BC060942.1 | - | 7 - 8 | 840 | 63 | |
Y17256.1 | - | 859 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |