Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131207.1 | NP_571282.1 | 4 - 5 | 871 | 62 | |
GenBank mRNA | AF001299.1 | - | 4 - 5 | 871 | 62 | |
BC162280.1 | - | 4 - 5 | 864 | 62 | ||
BC162303.1 | - | 4 - 5 | 864 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |