Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rarab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131399.1 | NP_571474.1 | 4 - 5 | 632 | 76 | |
XM_005164029.4 | XP_005164086.1 | 6 - 7 | 1450 | 76 | ||
GenBank mRNA | BC049301.1 | - | 4 - 5 | 632 | 76 | |
KF030797.1 | - | 4 - 5 | 979 | 76 | ||
L03399.1 | - | 4 - 5 | 801 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |