Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131565.1 | NP_571640.1 | 8 - 9 | 557 | 114 | |
XM_005163151.3 | XP_005163208.3 | 9 - 10 | 697 | 114 | ||
GenBank mRNA | AF180889.1 | - | 8 - 9 | 557 | 114 | |
BC053304.1 | - | 9 - 10 | 580 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |