Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200152.1 | NP_956446.1 | 3 - 4 | 542 | 79 | |
GenBank mRNA | BC044547.1 | - | 3 - 4 | 542 | 79 | |
BC066577.1 | - | 3 - 4 | 573 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |