Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4e2rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200759.2 | NP_957053.1 | 6 - 7 | 858 | 134 | |
XM_017353532.2 | XP_017209021.1 | 6 - 7 | 861 | 134 | ||
GenBank mRNA | BC059582.1 | - | 6 - 7 | 807 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |