Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205541.2 | NP_991104.2 | 1 - 2 | 228 | 83 | |
GenBank mRNA | BC045853.1 | - | 1 - 2 | 216 | 83 | |
BC065342.1 | - | 1 - 2 | 161 | 83 | ||
BC067379.1 | - | 1 - 2 | 155 | 83 | ||
BC164306.1 | - | 1 - 2 | 77 | 83 | ||
EH455821.1 | - | 1 - 2 | 195 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |