Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212968.2 | NP_998133.2 | 5 - 6 | 2257 | 103 | |
GenBank mRNA | AY507658.1 | - | 5 - 6 | 2258 | 103 | |
AY507659.1 | - | 5 - 6 | 2258 | 103 | ||
AY507660.1 | - | 5 - 6 | 2258 | 103 | ||
BC162109.1 | - | 5 - 6 | 2256 | 103 | ||
BC162480.1 | - | 5 - 6 | 2256 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |