Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123281.1 | NP_001116753.1 | 4 | 953 | 140 | |
GenBank mRNA | BC139546.1 | - | 4 | 945 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |