Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: six7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131354.2 | NP_571429.1 | 1 | 249 | 108 | |
GenBank mRNA | AF030282.1 | - | 1 | 385 | 108 | |
BC064701.1 | - | 1 | 249 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |