Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039513.1 | NP_001034602.1 | 6 | 947 | 100 | |
GenBank mRNA | AB201386.1 | - | 6 | 947 | 100 | |
BC121734.1 | - | 6 | 926 | 100 | ||
BC152092.1 | - | 6 | 942 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |