Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326493.1 | NP_001313422.1 | 4 | 1653 | 139 | |
GenBank mRNA | BC133081.1 | - | 4 | 1653 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |