Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spsb4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003474.1 | NP_001003474.1 | 1 | 252 | 144 | |
XM_005157796.4 | XP_005157853.1 | 3 | 637 | 144 | ||
XM_005157797.4 | XP_005157854.1 | 3 | 622 | 144 | ||
XM_009291907.3 | XP_009290182.1 | 4 | 656 | 144 | ||
XM_009291908.3 | XP_009290183.1 | 4 | 641 | 144 | ||
GenBank mRNA | BC078385.1 | - | 1 | 252 | 144 | |
BC155170.1 | - | 3 | 556 | 144 | ||
BC164637.1 | - | 1 | 233 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |