Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc85cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002628.1 | NP_001002628.1 | 7 | 2052 | 74 | |
XM_005160350.4 | XP_005160407.1 | 6 | 2208 | 74 | ||
GenBank mRNA | BC075952.1 | - | 7 | 2052 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |