Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syndig1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044781.1 | NP_001038246.1 | 3 | 643 | 59 | |
XM_005160575.4 | XP_005160632.1 | 4 | 1649 | 59 | ||
GenBank mRNA | BC171615.1 | - | 3 | 678 | 59 | |
BC171619.1 | - | 3 | 678 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |