Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: harbi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003734.1 | NP_001003734.1 | 2 - 3 | 717 | 82 | |
GenBank mRNA | BC078390.1 | - | 2 - 3 | 717 | 82 | |
BC100116.1 | - | 2 - 3 | 713 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |